Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS24
Entrez Gene ID
64951
Gene Name
mitochondrial ribosomal protein S24
Gene Aliases
HSPC335, MRP-S24, S24mt, bMRP-47, bMRP47
Chromosome Location
Chr.7: 43866558 - 43869546 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032014.2 NP_114403.1 1 - 2 87 107
GenBank mRNA BC012167.1 - 1 - 2 39 107
BC054865.1 - 1 - 2 54 107
CA413186.1 - 3 - 4 107

View Full Product Details