The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130848.2 | NP_570900.1 | 1 - 1 | 3094 | 74 |
GenBank mRNA | AB074498.1 | - | 1 - 1 | 3094 | 74 |