The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203373.2 | NP_976307.2 | 1 - 2 | 392 | 133 |
GenBank mRNA | BC065833.1 | - | 1 - 2 | 348 | 133 |