Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB18
Entrez Gene ID
401036
Gene Name
ankyrin repeat and SOCS box containing 18
Gene Aliases
ASB-18
Chromosome Location
Chr.2: 236194872 - 236264345 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212556.2 NP_997721.2 4 - 5 1106 116
GenBank mRNA AK123854.1 - 3 - 4 578 116

View Full Product Details