Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- SNU13
- Entrez Gene ID
-
4809
- Gene Name
- SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5)
- Gene Aliases
-
15.5K, FA-1, FA1, NHP2L1, NHPX, OTK27, SNRNP15-5, SPAG12, SSFA1
- Chromosome Location
-
Chr.22: 41673933 - 41690492 on Build GRCh38
- Gene Symbol
- ANXA2
- Entrez Gene ID
-
302
- Gene Name
- annexin A2
- Gene Aliases
-
ANX2, ANX2L4, CAL1H, HEL-S-270, LIP2, LPC2, LPC2D, P36, PAP-IV
- Chromosome Location
-
Chr.15: 60347151 - 60398025 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK309356.1 |
- |
- |
185 |
148 |
View Full Product Details