Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP5-7
Entrez Gene ID
440050
Gene Name
keratin associated protein 5-7
Gene Aliases
KRTAP5-3, KRTAP5.7
Chromosome Location
Chr.11: 71527267 - 71528164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012503.1 NP_001012521.1 1 - 1 663 111
GenBank mRNA AB126076.1 - 1 - 1 663 111
AJ628243.1 - 1 - 1 249 111
BC136967.1 - 1 - 1 652 111
BC136968.1 - 1 - 1 652 111

View Full Product Details