Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HCG27
Entrez Gene ID
253018
Gene Name
HLA complex group 27 (non-protein coding)
Gene Aliases
bCX101P6.9, bPG299F13.9, bQB115I13.2
Chromosome Location
Chr.6: 31197760 - 31203968 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026791.1 2 - 2 413 131
GenBank mRNA BC041465.2 - 1 - 1 292 131
BC104433.1 - 2 - 2 336 131

View Full Product Details