The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080518.1 | NP_001073987.1 | 4 - 5 | 530 | 65 |
XM_011540073.2 | XP_011538375.1 | 5 - 6 | 631 | 65 | |
XM_011540075.1 | XP_011538377.1 | 6 - 7 | 685 | 65 | |
XM_011540077.1 | XP_011538379.1 | 5 - 6 | 622 | 65 | |
XM_011540082.1 | XP_011538384.1 | 2 - 3 | 630 | 65 | |
XM_017016545.1 | XP_016872034.1 | 5 - 6 | 631 | 65 | |
XM_017016546.1 | XP_016872035.1 | 5 - 6 | 533 | 65 | |
XM_017016547.1 | XP_016872036.1 | 5 - 6 | 632 | 65 | |
XM_017016548.1 | XP_016872037.1 | 5 - 6 | 634 | 65 | |
GenBank mRNA | EF426482.1 | - | 4 - 5 | 530 | 65 |