Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP5-8
Entrez Gene ID
57830
Gene Name
keratin associated protein 5-8
Gene Aliases
KRTAP5-2, KRTAP5.8, UHSKerB
Chromosome Location
Chr.11: 71538025 - 71539207 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021046.2 NP_066384.2 1 - 1 961 102
GenBank mRNA AB126077.1 - 1 - 1 961 102
BX099510.1 - 1 - 1 296 102

View Full Product Details