The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004536.2 | NP_004527.2 | 2 - 3 | 581 | 87 |
NM_022892.1 | NP_075043.1 | 2 - 3 | 581 | 87 | |
GenBank mRNA | AB048534.1 | - | 2 - 3 | 581 | 87 |
U19251.1 | - | 2 - 3 | 154 | 87 |