Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHCHD2
Entrez Gene ID
51142
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Gene Aliases
C7orf17, MNRR1, NS2TP, PARK22
Chromosome Location
Chr.7: 56101562 - 56106630 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320327.1 NP_001307256.1 2 - 3 517 111
NM_016139.3 NP_057223.1 2 - 3 517 111
GenBank mRNA AA398444.1 - 2 - 3 448 111
AF078845.1 - 2 - 3 381 111
AI218619.1 - 2 - 3 450 111
AY605046.1 - 2 - 3 300 111
AY633613.1 - 2 - 3 321 111
BC003079.2 - 2 - 3 337 111
BC015639.2 - 2 - 3 321 111
BC066331.1 - 2 - 3 340 111
BC071985.1 - 2 - 3 337 111
BC100275.1 - 2 - 3 333 111
BF347122.1 - - 392 111
CN363208.1 - 2 - 3 357 111

View Full Product Details