The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142776.1 | NP_001136248.1 | 2 - 3 | 408 | 93 |
NM_024111.3 | NP_077016.2 | 2 - 3 | 408 | 93 | |
XM_017022563.1 | XP_016878052.1 | 3 - 4 | 4932 | 93 | |
XM_017022564.1 | XP_016878053.1 | 4 - 5 | 2714 | 93 | |
XM_017022565.1 | XP_016878054.1 | 3 - 4 | 4921 | 93 | |
GenBank mRNA | AY702027.1 | - | 2 - 3 | 270 | 93 |
BC001683.1 | - | 2 - 3 | 362 | 93 | |
BC001847.2 | - | 2 - 3 | 353 | 93 | |
BC019625.1 | - | 2 - 3 | 364 | 93 | |
BQ900544.1 | - | 2 - 3 | 408 | 93 |