Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHDC3
Entrez Gene ID
116138
Gene Name
kelch domain containing 3
Gene Aliases
PEAS, dJ20C7.3
Chromosome Location
Chr.6: 43014103 - 43021298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057161.3 NP_476502.1 7 - 8 1124 152
NR_040101.1 6 - 7 1008 152
GenBank mRNA AB055925.1 - 7 - 8 988 152
AK097719.1 - 5 - 6 850 152
AK290384.1 - 7 - 8 980 152
AK308917.1 - 7 - 8 1010 152
BC000295.2 - 7 - 8 944 152
BC001789.1 - 7 - 8 959 152
BC001793.2 - 7 - 8 922 152
BC007296.1 - 7 - 8 974 152
BC009460.1 - 7 - 8 972 152
BC021546.1 - 7 - 8 961 152
BC041793.1 - 6 - 7 862 152
BC045612.1 - 6 - 7 862 152

View Full Product Details