The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198896.1 | NP_942599.1 | 3 - 4 | 704 | 151 |
GenBank mRNA | AF130986.1 | - | 3 - 4 | 183 | 151 |
AF498939.1 | - | 3 - 4 | 183 | 151 | |
AK289748.1 | - | 3 - 4 | 622 | 151 | |
AK290132.1 | - | 3 - 4 | 353 | 151 | |
BC096818.1 | - | 3 - 4 | 638 | 151 | |
BI552973.1 | - | 3 - 4 | 639 | 152 | |
BQ422263.1 | - | 3 - 4 | 186 | 151 | |
BT019698.1 | - | 3 - 4 | 183 | 151 | |
M28212.1 | - | 3 - 4 | 253 | 151 | |
X13904.1 | - | 3 - 4 | 406 | 151 |