The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000979.3 | NP_000970.1 | 2 - 3 | 369 | 114 |
GenBank mRNA | AK026558.1 | - | 2 - 3 | 127 | 114 |
AK293386.1 | - | 2 - 3 | 128 | 114 | |
BC000374.2 | - | 1 - 2 | 101 | 114 | |
BC009708.2 | - | 2 - 3 | 115 | 114 | |
BC021743.2 | - | 2 - 3 | 127 | 114 | |
BM764512.1 | - | 2 - 3 | 487 | 114 | |
L11566.1 | - | 2 - 3 | 109 | 114 |