The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000979.3 | NP_000970.1 | 5 - 6 | 696 | 75 |
NM_001270490.1 | NP_001257419.1 | 4 - 5 | 609 | 75 | |
NR_073022.1 | 5 - 6 | 688 | 75 | ||
GenBank mRNA | AI921565.1 | - | 2 - 3 | 75 | |
AK026558.1 | - | 5 - 6 | 454 | 75 | |
BC000374.2 | - | 4 - 5 | 428 | 75 | |
BC009708.2 | - | 5 - 6 | 442 | 75 | |
BC021743.2 | - | 5 - 6 | 454 | 75 | |
CD246847.1 | - | 4 - 5 | 393 | 75 | |
CD389415.1 | - | 5 - 6 | 460 | 75 | |
DQ403032.1 | - | 4 - 5 | 352 | 75 | |
L11566.1 | - | 5 - 6 | 436 | 75 |