Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST1H2AC
Entrez Gene ID
8334
Gene Name
histone cluster 1, H2ac
Gene Aliases
H2A/l, H2AFL, dJ221C16.4
Chromosome Location
Chr.6: 26124145 - 26124690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003512.3 NP_003503.1 1 - 1 505 66
GenBank mRNA BC017379.1 - 1 - 1 469 66
BC050602.1 - 1 - 1 461 66
BC085010.1 - 1 - 1 466 66
U90551.1 - 1 - 1 514 66

View Full Product Details