Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YOD1
Entrez Gene ID
55432
Gene Name
YOD1 deubiquitinase
Gene Aliases
DUBA8, OTUD2, PRO0907
Chromosome Location
Chr.1: 207043849 - 207052980 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276320.1 NP_001263249.1 3 - 4 416 159
NM_018566.3 NP_061036.3 1 - 2 390 159
GenBank mRNA AK128014.1 - 3 - 4 416 159
AL833081.2 - 1 - 2 322 159
BC137166.1 - 1 - 2 382 159
BC137167.1 - 1 - 2 382 159

View Full Product Details