Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL6
Entrez Gene ID
6128
Gene Name
ribosomal protein L6
Gene Aliases
L6, SHUJUN-2, TAXREB107, TXREB1
Chromosome Location
Chr.12: 112405181 - 112418850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000970.4 NP_000961.2 6 - 7 773 82
NM_001024662.2 NP_001019833.1 6 - 7 949 82
NM_001320137.1 NP_001307066.1 6 - 7 954 82
NM_001320138.1 NP_001307067.1 6 - 7 988 82
NM_001320139.1 NP_001307068.1 6 - 7 835 82
NM_001320140.1 NP_001307069.1 6 - 7 997 82
NM_001320141.1 NP_001307070.1 8 - 9 1062 82
NM_001320142.1 NP_001307071.1 6 - 7 709 82
XM_017019781.1 XP_016875270.1 8 - 9 2097 82
XM_017019782.1 XP_016875271.1 8 - 9 1258 82
GenBank mRNA AF261087.1 - 1 - 1 743 82
AI583261.1 - 1 - 1 50 82
AK298117.1 - 4 - 5 475 82
AK299470.1 - 7 - 8 703 82
AK311863.1 - 6 - 7 737 82
AY320410.1 - 3 - 4 293 82
BC004138.2 - 1 - 1 749 82
BC013863.1 - 1 - 1 723 82
BC020679.1 - 1 - 1 737 82
BC022444.1 - 6 - 7 904 82
BC031009.1 - 1 - 1 737 82
BC032299.1 - 1 - 1 734 82
BC071912.1 - 1 - 1 726 82
BC104824.1 - 6 - 7 749 82
BC104826.1 - 6 - 7 749 82
BG030827.1 - 1 - 1 499 82
D17554.1 - 6 - 7 739 82
X69391.1 - 6 - 7 744 82

Gene Symbol
RPL6P27
Entrez Gene ID
645387
Gene Name
ribosomal protein L6 pseudogene 27
Gene Aliases
RPL6_10_1582
Chromosome Location
Chr.18: 6462110 - 6463029 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK126570.1 - - 1485 82

View Full Product Details