Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSG6
Entrez Gene ID
5675
Gene Name
pregnancy specific beta-1-glycoprotein 6
Gene Aliases
PSBG-10, PSBG-12, PSBG-6, PSG10, PSGGB
Chromosome Location
Chr.19: 42902082 - 42917924 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002782.4 NP_002773.1 5 - 6 1379 109
GenBank mRNA M33666.1 - 5 - 6 1299 109

View Full Product Details