Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMM2
Entrez Gene ID
5373
Gene Name
phosphomannomutase 2
Gene Aliases
CDG1, CDG1a, CDGS, PMI, PMI1, PMM 2
Chromosome Location
Chr.16: 8797813 - 8849337 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000303.2 NP_000294.1 1 - 2 139 157
XM_005255374.4 XP_005255431.1 1 - 2 139 157
GenBank mRNA AK291537.1 - 1 - 2 110 157
AK296123.1 - 1 - 2 132 157
AK296630.1 - 1 - 2 85 157
AK299979.1 - 1 - 2 110 157
AK300785.1 - 1 - 2 115 157
AK304320.1 - 1 - 2 102 157
AK311035.1 - 1 - 2 110 157
AK312532.1 - 1 - 2 110 157
BC008310.1 - 1 - 2 81 157
DC366756.1 - 1 - 2 146 157
KU178265.1 - 1 - 2 69 157
KU178266.1 - 1 - 2 69 157
U85773.1 - 1 - 2 117 157

View Full Product Details