The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138964.2 | NP_620414.1 | 1 - 2 | 478 | 106 |
GenBank mRNA | AB084080.1 | - | 1 - 2 | 478 | 106 |
AF506287.1 | - | 1 - 2 | 478 | 106 | |
AY089976.1 | - | 1 - 2 | 478 | 106 | |
BC100960.2 | - | 1 - 2 | 478 | 106 | |
EF577399.1 | - | 1 - 2 | 478 | 106 |