The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181600.1 | NP_853631.1 | 1 - 1 | 430 | 75 |
GenBank mRNA | AB096941.1 | - | 1 - 1 | 452 | 75 |
BC131691.1 | - | 1 - 1 | 434 | 75 |