Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IRGC
Entrez Gene ID
56269
Gene Name
immunity related GTPase cinema
Gene Aliases
CINEMA, IFGGE, IRGC1, Iigp5, R30953_1
Chromosome Location
Chr.19: 43716062 - 43720021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019612.3 NP_062558.1 2 - 2 1371 111
XM_005259058.3 XP_005259115.1 2 - 2 1465 111
XM_006723283.3 XP_006723346.1 2 - 2 1402 111
XM_011527118.2 XP_011525420.1 3 - 3 1405 111
GenBank mRNA BC066939.1 - 2 - 2 1347 111

View Full Product Details