The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040061.2 | NP_001035150.1 | 2 - 3 | 354 | 56 |
XM_005247443.3 | XP_005247500.1 | 3 - 4 | 440 | 56 | |
GenBank mRNA | AK125319.1 | - | 2 - 3 | 354 | 56 |