The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102654.1 | NP_001096124.1 | 2 - 2 | 794 | 107 |
NM_002527.4 | NP_002518.1 | 1 - 1 | 627 | 107 | |
XM_011520963.2 | XP_011519265.1 | 2 - 2 | 838 | 107 | |
GenBank mRNA | AK293895.1 | - | 2 - 2 | 798 | 107 |
BC069773.1 | - | 1 - 1 | 627 | 107 | |
BC107075.1 | - | 1 - 1 | 107 | ||
CR541906.1 | - | 1 - 1 | 544 | 107 | |
X53655.1 | - | 2 - 2 | 587 | 107 |