Assay Design
The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.
Gene Transcripts
- Gene Symbol
- CTRB1
- Entrez Gene ID
-
1504
- Gene Name
- chymotrypsinogen B1
- Gene Aliases
-
CTRB
- Chromosome Location
-
Chr.16: 75218986 - 75224924 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AA844834.1 |
- |
1 - 1 |
|
104 |
AW338596.1 |
- |
1 - 1 |
|
104 |
M24400.1 |
- |
6 - 6 |
792 |
104 |
- Gene Symbol
- CTRB2
- Entrez Gene ID
-
440387
- Gene Name
- chymotrypsinogen B2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.16: 75204096 - 75207174 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK124403.1 |
- |
4 - 4 |
2208 |
104 |
View Full Product Details