Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HOXB3
- Entrez Gene ID
-
3213
- Gene Name
- homeobox B3
- Gene Aliases
-
HOX2, HOX2G, Hox-2.7
- Chromosome Location
-
Chr.17: 48548870 - 48590369 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK000839.1 |
- |
1 - 1 |
151 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 10a |
MIR10A |
MIRN10A, hsa-mir-10a, miRNA10A, mir-10a |
-370.0
|
Sense |
Chr. - 48579838 - 48579947 on Build GRCh38 |
Downstream |
HOXB cluster antisense RNA 3 |
HOXB-AS3 |
|
8.2kb
|
Anti-sense |
Chr. - 48590420 - 48606414 on Build GRCh38 |
View Full Product Details