Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPPH1
- Entrez Gene ID
-
85495
- Gene Name
- ribonuclease P RNA component H1
- Gene Aliases
-
H1RNA, RPPH1-1
- Chromosome Location
-
Chr.14: 20343071 - 20343411 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002312.1 |
|
1 - 1 |
254 |
68 |
GenBank mRNA |
CA413366.1 |
- |
1 - 1 |
|
68 |
CV309337.1 |
- |
1 - 1 |
172 |
68 |
X15624.1 |
- |
1 - 1 |
253 |
68 |
- Gene Symbol
- PARP2
- Entrez Gene ID
-
10038
- Gene Name
- poly(ADP-ribose) polymerase 2
- Gene Aliases
-
ADPRT2, ADPRTL2, ADPRTL3, ARTD2, PARP-2, pADPRT-2
- Chromosome Location
-
Chr.14: 20343041 - 20357904 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CA413345.1 |
- |
1 |
|
|
Chr. 14 - 20343049 - 20343685 [+] on Build GRCh38 |
68 |
GenBankmRNA |
CV427054.1 |
- |
1 |
169 |
|
Chr. 14 - 20343096 - 20343326 [-] on Build GRCh38 |
68 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cyclin B1 interacting protein 1 |
CCNB1IP1 |
C14orf18, HEI10 |
-9.8kb
|
Sense |
Chr. - 20311368 - 20333312 on Build GRCh38 |
View Full Product Details