Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORD51
- Entrez Gene ID
-
26798
- Gene Name
- small nucleolar RNA, C/D box 51
- Gene Aliases
-
RNU51, U51
- Chromosome Location
-
Chr.2: 206161881 - 206161950 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002589.1 |
|
1 - 1 |
36 |
70 |
GenBank mRNA |
X96650.1 |
- |
1 - 1 |
36 |
70 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA371392.1 |
- |
1 |
87 |
|
Chr. 2 - 206161830 - 206162106 [+] on Build GRCh38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NADH:ubiquinone oxidoreductase core subunit S1 |
NDUFS1 |
CI-75Kd, CI-75k, PRO1304 |
-2.4kb
|
Anti-sense |
Chr. - 206121971 - 206159519 on Build GRCh38 |
Downstream |
small nucleolar RNA, H/ACA box 41 |
SNORA41 |
ACA41, SNORA41A |
278.0
|
Sense |
Chr. - 206162228 - 206162359 on Build GRCh38 |
View Full Product Details