Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PPP4R3CP
- Entrez Gene ID
-
139420
- Gene Name
- protein phosphatase 4 regulatory subunit 3C, pseudogene
- Gene Aliases
-
FLFL3P, SMEK3P, smk1
- Chromosome Location
-
Chr.X: 27460211 - 27463341 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002784.2 |
|
1 - 1 |
2256 |
113 |
GenBank mRNA |
AK131475.1 |
- |
1 - 1 |
2256 |
113 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105373150 |
LOC105373150 |
|
-61.2kb
|
Sense |
Chr. - 27153368 - 27399005 on Build GRCh38 |
Downstream |
DDB1 and CUL4 associated factor 8 like 2 |
DCAF8L2 |
WDR42C |
283.5kb
|
Anti-sense |
Chr. - 27746809 - 27748821 on Build GRCh38 |
View Full Product Details