Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHF2P1
Entrez Gene ID
266695
Gene Name
PHD finger protein 2 pseudogene 1
Gene Aliases
DKFZp686A1627
Chromosome Location
Chr.13: 19045351 - 19072391 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AL832753.1 - 10 - 11 3677 88


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 442 LINC00442 -33.5kb Anti-sense Chr. - 19008259 - 19012634 on Build GRCh38
Downstream tubulin alpha 3c TUBA3C TUBA2, bA408E5.3 125.5kb Sense Chr. - 19173770 - 19181852 on Build GRCh38

View Full Product Details