Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KC6
Entrez Gene ID
641516
Gene Name
keratoconus gene 6
Gene Aliases
-
Chromosome Location
Chr.18: 41480271 - 41520597 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_002838.1 3 - 4 511 133
GenBank mRNA AY762618.1 - 3 - 4 511 133

View Full Product Details