Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORD19
- Entrez Gene ID
-
692089
- Gene Name
- small nucleolar RNA, C/D box 19
- Gene Aliases
-
HBII-108
- Chromosome Location
-
Chr.3: 52689240 - 52689315 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003047.1 |
|
1 - 1 |
33 |
70 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI248847.1 |
- |
1 |
|
|
Chr. 3 - 52689080 - 52689618 [-] on Build GRCh38 |
70 |
GenBankmRNA |
BE842778.1 |
- |
1 |
|
|
Chr. 3 - 52689219 - 52689483 [-] on Build GRCh38 |
70 |
GenBankmRNA |
BX478610.1 |
- |
1 |
68 |
|
Chr. 3 - 52689205 - 52689776 [+] on Build GRCh38 |
70 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
polybromo 1 |
PBRM1 |
BAF180, PB1 |
-3.3kb
|
Anti-sense |
Chr. - 52545352 - 52685933 on Build GRCh38 |
Downstream |
small nucleolar RNA, C/D box 19B |
SNORD19B |
|
1.4kb
|
Sense |
Chr. - 52690738 - 52690830 on Build GRCh38 |
View Full Product Details