Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ITPK1-AS1
- Entrez Gene ID
-
319085
- Gene Name
- ITPK1 antisense RNA 1
- Gene Aliases
-
C14orf85, ITPK1-AS, ITPK1AS, NCRNA00203
- Chromosome Location
-
Chr.14: 93067452 - 93072152 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002808.2 |
|
1 - 1 |
4245 |
107 |
GenBank mRNA |
AK024887.1 |
- |
1 - 1 |
4246 |
107 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
inositol-tetrakisphosphate 1-kinase |
ITPK1 |
ITRPK1 |
|
Anti-sense |
Chr. - 92936914 - 93115918 on Build GRCh38 |
Downstream |
modulator of apoptosis 1 |
MOAP1 |
MAP-1, PNMA4 |
110.0kb
|
Anti-sense |
Chr. - 93182196 - 93184904 on Build GRCh38 |
View Full Product Details