The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001093730.1 | NP_001087199.1 | 4 - 5 | 494 | 82 |
GenBank mRNA | DQ516347.1 | - | 4 - 5 | 494 | 82 |