The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114133.2 | NP_001107605.1 | 1 - 2 | 293 | 104 |
GenBank mRNA | AB188489.1 | - | 1 - 2 | 262 | 104 |
AK095931.1 | - | 1 - 2 | 262 | 104 | |
AK295873.1 | - | 1 - 2 | 182 | 104 | |
DC391557.1 | - | 1 - 2 | 293 | 104 |