Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4F3
Entrez Gene ID
26683
Gene Name
olfactory receptor family 4 subfamily F member 3
Gene Aliases
-
Chromosome Location
Chr.5: 181342142 - 181370030 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005224.1 NP_001005224.1 1 - 1 648 126
XM_017009338.1 XP_016864827.1 2 - 2 4865 126
GenBank mRNA BC137547.1 - 1 - 1 667 126
BC137568.1 - 1 - 1 667 126

Gene Symbol
OR4F16
Entrez Gene ID
81399
Gene Name
olfactory receptor family 4 subfamily F member 16
Gene Aliases
OR1-1, OR7-21
Chromosome Location
Chr.1: 683910 - 720118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005277.1 NP_001005277.1 1 - 1 648 126
XM_017002408.1 XP_016857897.1 3 - 3 3380 126
XM_017002409.1 XP_016857898.1 4 - 4 3298 126
XM_017002410.1 XP_016857899.1 2 - 2 3162 126
XM_017002411.1 XP_016857900.1 2 - 2 3151 126

Gene Symbol
OR4F21
Entrez Gene ID
441308
Gene Name
olfactory receptor family 4 subfamily F member 21
Gene Aliases
OR4F21P
Chromosome Location
Chr.8: 166086 - 167024 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005504.1 NP_001005504.1 1 - 1 648 126

Gene Symbol
OR4F29
Entrez Gene ID
729759
Gene Name
olfactory receptor family 4 subfamily F member 29
Gene Aliases
OR7-21
Chromosome Location
Chr.1: 450740 - 451678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005221.2 NP_001005221.2 1 - 1 648 126

View Full Product Details