Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
HY169091.1 |
- |
1 - 2 |
128 |
|
Chr. 7 - 27187894 - 27188992 [+] on Build GRCh38 |
116 |
GenBankmRNA |
BG746816.1 |
- |
1 - 2 |
204 |
|
Chr. 7 - 27187819 - 27189222 [+] on Build GRCh38 |
116 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
HOXA11 antisense RNA |
HOXA11-AS |
HOXA-AS5, HOXA11-AS1, HOXA11AS, HOXA11S, NCRNA00076 |
|
Sense |
Chr. - 27185408 - 27189293 on Build GRCh38 |
Downstream |
homeobox A13 |
HOXA13 |
HOX1, HOX1J |
4.2kb
|
Anti-sense |
Chr. - 27196880 - 27200106 on Build GRCh38 |
View Full Product Details