Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST7-OT3
Entrez Gene ID
93655
Gene Name
ST7 overlapping transcript 3
Gene Aliases
NCRNA00026, ST7, ST7OT3
Chromosome Location
Chr.7: 117182681 - 117209937 on Build GRCh38
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location Amplicon Length
RefSeq NR_002332.2
6 - 7 579 95
GenBank mRNA AF400044.1 - LIT1900
ST7OT3
6 - 7 579 95


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream ST7 antisense RNA 2 ST7-AS2 ST7AS2, ST7OT2 -46.0kb Anti-sense Chr. - 117072072 - 117145592 on Build GRCh38
Downstream Wnt family member 2 WNT2 INT1L1, IRP 78.1kb Anti-sense Chr. - 117276631 - 117323289 on Build GRCh38

View Full Product Details