The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NR_003141.3 | 4 - 4 | 630 | 132 | ||
GenBank mRNA | AJ224167.1 | - | LIT1319 U19HG |
4 - 4 | 251 | 132 |
CN337312.1 | - | 2 - 2 | 196 | 132 | ||
DA203569.1 | - | 1 - 1 | 114 | 132 | ||
DB192707.1 | - | 4 - 4 | 293 | 132 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | small nucleolar RNA, H/ACA box 74A | SNORA74A | RNU19, U19 | -72.0 | Sense | Chr. - 139278780 - 139278979 on Build GRCh38 |
Downstream | poly(A) binding protein interacting protein 2 | PAIP2 | PAIP-2, PAIP2A | 62.2kb | Sense | Chr. - 139341832 - 139369720 on Build GRCh38 |