Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI671175.1 - 1 248 Chr. 6 - 26086291 - 26086736 [+] on Build GRCh38 105
GenBankmRNA DB311093.1 - 1 250 Chr. 6 - 26086291 - 26086810 [+] on Build GRCh38 105
GenBankmRNA AK123489.1 - 1 Chr. 6 - 26086291 - 26088155 [-] on Build GRCh38 105
GenBankmRNA AI916326.1 - 1 256 Chr. 6 - 26086290 - 26086693 [+] on Build GRCh38 105


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream histone cluster 1, H1c HIST1H1C H1.2, H1C, H1F2, H1s-1 -29.8kb Anti-sense Chr. - 26055740 - 26056471 on Build GRCh38
Downstream hemochromatosis HFE HFE1, HH, HLA-H, MVCD7, TFQTL2 545.0 Sense Chr. - 26087281 - 26096216 on Build GRCh38

View Full Product Details