Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2F6
Entrez Gene ID
1876
Gene Name
E2F transcription factor 6
Gene Aliases
E2F-6
Chromosome Location
Chr.2: 11444375 - 11466177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278276.1 NP_001265205.1 3 - 4 599 74
NM_001278277.1 NP_001265206.1 4 - 5 670 74
GenBank mRNA AK096197.1 - 4 - 5 372 74
BU076470.1 - 3 - 4 277 74


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA642382.1 - 3 - 4 578 Chr. 2 - 11453746 - 11466156 [-] on Build GRCh38 74
GenBankmRNA BX380302.2 - 3 - 4 568 Chr. 2 - 11451658 - 11466146 [-] on Build GRCh38 74
GenBankmRNA DA658899.1 - 4 - 5 373 Chr. 2 - 11453614 - 11465879 [-] on Build GRCh38 74


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 570 LINC00570 ncRNA-a5 -50.5kb Anti-sense Chr. - 11393981 - 11403077 on Build GRCh38
Downstream growth regulation by estrogen in breast cancer 1 GREB1 78.6kb Anti-sense Chr. - 11534116 - 11642788 on Build GRCh38

View Full Product Details