The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_002216.1 | 2 - 3 | 319 | 139 | |
GenBank mRNA | AK124700.1 | - | 2 - 3 | 319 | 139 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | thymosin beta 15a | TMSB15A | TMSB15, TMSB15B, TMSL8, TMSNB, Tb15, TbNB | -44.7kb | Anti-sense | Chr. - 102513682 - 102516771 on Build GRCh38 |
Downstream | armadillo repeat containing, X-linked 5 | ARMCX5 | GASP5 | 37.2kb | Sense | Chr. - 102599168 - 102604159 on Build GRCh38 |