Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SNORD59A
- Entrez Gene ID
-
26789
- Gene Name
- small nucleolar RNA, C/D box 59A
- Gene Aliases
-
RNU59, U59
- Chromosome Location
-
Chr.12: 56645027 - 56645101 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002737.1 |
|
1 - 1 |
39 |
75 |
GenBank mRNA |
X96659.1 |
- |
1 - 1 |
39 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small nucleolar RNA, C/D box 59B |
SNORD59B |
U59B |
-1.3kb
|
Sense |
Chr. - 56643680 - 56643754 on Build GRCh38 |
Downstream |
prostaglandin E synthase 3 |
PTGES3 |
P23, TEBP, cPGES |
18.2kb
|
Sense |
Chr. - 56663341 - 56689250 on Build GRCh38 |
View Full Product Details