Product Details

Assay Reference Genome
Location

Chr.7:44157502 on build GRCh38
Cytoband
7p13
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
2645
Gene Symbol
GCK
Gene Name
glucokinase
Gene Aliases
FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2
Location
Chr.7:44143213-44189439 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
GCK NM_000162.3 NP_000153.1
NM_033507.1 NP_277042.1
NM_033508.1 NP_277043.1
XM_017011966.1 XP_016867455.1
AK122876.1
BC001890.1 AAH01890.1
BT007062.1 AAP35711.1
DA640823.1
EU008605.1
M69051.1 AAB59563.1
M88011.1 AAA51824.1
M90299.1 AAA52562.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv830983 Chr7:44068842 - 44201624 on Build GRCh38 Loss POLM GCK MIR4649 AEBP1 MIR6838 YKT6 MYL7 POLD2
nsv8082 Chr7:43956156 - 44207229 on Build GRCh38 Gain RASA4CP POLM GCK AEBP1 DBNL MYL7 POLD2 POLR2J4 MIR4649 PGAM2 MIR6838 SPDYE1 YKT6 LINC00957 MIR6837
nsv428165 Chr7:43855828 - 44172960 on Build GRCh38 Gain RASA4CP POLM GCK AEBP1 LOC107986791 DBNL MYL7 UBE2D4 POLD2 POLR2J4 MIR4649 PGAM2 URGCP-MRPS24 MIR6838 SPDYE1 MRPS24 URGCP LINC00957 MIR6837
nsv509195 Chr7:44147118 - 44261782 on Build GRCh38 Insertion GCK YKT6 CAMK2B

View Full Product Details