Product Details

Assay Reference Genome
Location

Chr.7:100409947 on build GRCh38
Cytoband
7q22.1
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
55063
Gene Symbol
ZCWPW1
Gene Name
zinc finger CW-type and PWWP domain containing 1
Gene Aliases
ZCW1
Location
Chr.7:100400826-100429239 on build GRCh38
Assay Gene Location
Within Intron 9
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
ZCWPW1 NM_001258008.1 NP_001244937.1
NM_017984.4 NP_060454.3
XM_005250479.2 XP_005250536.1
XM_005250480.2 XP_005250537.1
XM_006716035.3 XP_006716098.1
XM_006716036.2 XP_006716099.1
XM_006716037.3 XP_006716100.1
XM_006716038.3 XP_006716101.1
XM_006716040.3 XP_006716103.1
XM_017012379.1 XP_016867868.1
XM_017012380.1 XP_016867869.1
AK000919.1 BAA91424.1
AK093038.1 BAC04028.1
AK300745.1
AK302108.1
AL136735.1 CAB66669.1
BC002725.1 AAH02725.1
KU178677.1
KU178678.1
KU178679.1
KU178680.1
KU178681.1
KU178682.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2734888 Chr7:99975623 - 101290285 on Build GRCh38 Deletion ZASP AZGP1 CLDN15 TRIP6 EPO GPC2 POP7 NAT16 ACHE PCOLCE-AS1 GIGYF1 MOSPD3 MUC12 MIR25 EPHB4 AZGP1P1 PILRB C7orf61 LOC102724094 SAP25 PLOD3 PCOLCE STAG3 AGFG2 UFSP1 CNPY4 PMS2P1 SLC12A9 PPP1R35 SPDYE3 SRRT VGF STAG3L5P-PVRIG2P-PILRB ZCWPW1 ZAN MEPCE TSC22D4 MIR106B COPS6 LAMTOR4 MCM7 MIR93 LOC105375429 C7orf43 FIS1 ZNF3 TRIM56 FBXO24 NYAP1 LRCH4 PILRA ZKSCAN1 MIR6875 GATS ACTL6B MIR4653 MUC3A MUC17 MIR6840 ZSCAN21 PVRIG2P STAG3L5P MBLAC1 AP1S1 MOGAT3 ZNHIT1 AP4M1 GAL3ST4 SERPINE1 MIR4658 TFR2 TAF6 GNB2 PVRIG
nsv831071 Chr7:100320402 - 100515790 on Build GRCh38 Loss MEPCE TSC22D4 PMS2P1 MIR6840 PVRIG2P STAG3L5P PPP1R35 SPDYE3 NYAP1 PILRA PILRB STAG3L5P-PVRIG2P-PILRB C7orf61 ZCWPW1

View Full Product Details