Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOLGA8DP
Entrez Gene ID
100132979
Gene Name
golgin A8 family member D, pseudogene
Gene Aliases
GOLGA8D
Chromosome Location
Chr.15: 23157340 - 23170783 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027407.1 3 - 4 354 68
GenBank mRNA AK292534.1 - 3 - 4 240 68
BC110417.1 - 3 - 4 325 68

Gene Symbol
GOLGA8CP
Entrez Gene ID
729786
Gene Name
golgin A8 family member C, pseudogene
Gene Aliases
GOLGA8C
Chromosome Location
Chr.15: 20562349 - 20575703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027411.2 3 - 4 354 68
GenBank mRNA DB085317.1 - 3 - 4 328 68

Gene Symbol
GOLGA8EP
Entrez Gene ID
390535
Gene Name
golgin A8 family member E, pseudogene
Gene Aliases
GOLGA8E
Chromosome Location
Chr.15: 22424673 - 22438026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033350.1 3 - 4 354 68
GenBank mRNA AK310240.1 - 3 - 4 328 68
BC063309.1 - 3 - 4 328 68
DQ309037.1 - 3 - 4 328 68

Gene Symbol
GOLGA8F
Entrez Gene ID
100132565
Gene Name
golgin A8 family member F
Gene Aliases
-
Chromosome Location
Chr.15: 28378638 - 28391266 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033351.1 3 - 4 354 68
GenBank mRNA AK057241.1 - 3 - 4 273 68
AK302625.1 - 3 - 4 328 68
BC144672.1 - 1 - 2 51 68
BC150302.1 - 3 - 4 328 68
DB071191.1 - 3 - 4 354 68
DQ309038.1 - 3 - 4 243 68

Gene Symbol
GOLGA8G
Entrez Gene ID
283768
Gene Name
golgin A8 family member G
Gene Aliases
-
Chromosome Location
Chr.15: 28519611 - 28532997 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033353.1 3 - 4 354 68
GenBank mRNA AK302813.1 - 3 - 4 348 68
BC066774.1 - 3 - 4 328 68
BC099913.1 - 3 - 4 327 68
BC125193.1 - 2 - 3 146 68
BC125194.1 - 2 - 3 146 68

Gene Symbol
GOLGA8S
Entrez Gene ID
653061
Gene Name
golgin A8 family member S
Gene Aliases
-
Chromosome Location
Chr.15: 23354748 - 23368324 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_038843.1 3 - 4 328 68
GenBank mRNA DB053404.1 - 3 - 4 328 68

View Full Product Details