The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126057.2 | NP_001119529.2 | 10 - 11 | 1353 | 91 |
NM_001126058.2 | NP_001119530.2 | 9 - 10 | 1317 | 91 | |
NM_001190348.1 | NP_001177277.1 | 12 - 13 | 1467 | 91 | |
NM_001190349.1 | NP_001177278.1 | 8 - 9 | 1266 | 91 | |
GenBank mRNA | AK125695.1 | - | 2 - 3 | 2668 | 91 |
AY358412.1 | - | 12 - 13 | 1479 | 91 | |
AY789696.1 | - | 9 - 10 | 1140 | 91 | |
AY789697.1 | - | 10 - 11 | 1176 | 91 | |
EF158324.1 | - | 3 - 4 | 423 | 91 | |
EF158325.1 | - | 4 - 5 | 297 | 91 | |
EF158326.1 | - | 7 - 8 | 438 | 91 |