Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- DMKN
- Entrez Gene ID
-
93099
- Gene Name
- dermokine
- Gene Aliases
-
UNQ729, ZD52F10
- Chromosome Location
-
Chr.19: 35497217 - 35513678 on Build GRCh38
View Full Product Details